Bis(phenylacetyl)disulfide 97%

Supplier: Ambeed
Warning

Synonyms: Phenylacetyl Disulphide, 2-Phenylacetic dithioperoxyanhydride

A277777-100G A277777-10G A277777-1G A277777-500G A277777-25G A277777-5G
103853-302EA 11 USD
103853-302 76723-170 76723-172 76723-174 76723-176 77093-204 77093-206
Bis(phenylacetyl)disulfide 97%
Bis(phenylacetyl)disulfide

Precaución: for Research Use Only

Formula: C₁₆H₁₄O₂S₂
MW: 302.41 g/mol
Boiling Pt: 481.1 °C (760 mm Hg)
Melting Pt: 59…63 °C
Flash Pt: 210.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00513572
CAS Number: 15088-78-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR