2-Phenyl-9H-carbazole 97%

Supplier: Ambeed
Warning

A447258-1G A447258-25G A447258-5G
103827-760EA 9 USD
103827-760 103838-828 76842-626 76842-628 76842-630
2-Phenyl-9H-carbazole 97%
2-Phenyl-9H-carbazole

Precaución: for Research Use Only

Formula: C₁₈H₁₃N
Melting Pt: 285…289 °C
Storage Temperature: Ambient
MDL Number: MFCD26402145
CAS Number: 88590-00-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR