2-Phenylindole 98%
Supplier: Ambeed
|
Warning
|
A190721-100G
A190721-10G
A190721-500G
A190721-25G
A190721-1000G
103809-246EA
94
USD
103809-246
76873-036
76873-038
76873-040
77084-744
77302-232
2-Phenylindole 98%
2-Phenylindole
Precaución: for Research Use Only
Formula:
C₁₄H₁₁N Melting Pt: 188…190 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005608 CAS Number: 948-65-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...