2-Phenylbenzimidazole 98%

Supplier: Ambeed
Warning

Synonyms: 2-Phenyl-1H-benzimidazole, 2-Phenyl-1H-benzo[d]imidazole

A192102-100G A192102-10G A192102-25G A192102-5G
103847-022EA 5 USD
103847-022 76690-924 76690-926 77084-898 77084-900
2-Phenylbenzimidazole 98%
2-Phenylbenzimidazole

Precaución: for Research Use Only

Formula: C₁₃H₁₀N₂
MW: 194.24 g/mol
Melting Pt: 293…296 °C
Flash Pt: 210 °C
Storage Temperature: Ambient
MDL Number: MFCD00005592
CAS Number: 716-79-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR