2-Phenyl-1,3-dithiane 95%

Supplier: Ambeed
Warning

A410559-25G A410559-5G A410559-100G A410559-10G A410559-1G
103827-216EA 5 USD
103827-216 103838-264 76841-786 76841-788 77278-866 77278-868 77307-148
2-Phenyl-1,3-dithiane 95%
2-Phenyl-1,3-dithiane

Precaución: for Research Use Only

Formula: C₁₀H₁₂S₂
MW: 196.34 g/mol
Boiling Pt: 326.8 °C (760 mm Hg)
Flash Pt: 158.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00006656
CAS Number: 5425-44-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR