(2-Aminoethoxy)benzene 98%

Supplier: Ambeed
Danger

Synonyms: ω-Aminophenetole, 2-Phenoxyethylamine, 2-Aminoethyl phenyl ether, β-Aminophenetole

A262607-100G A262607-25G A262607-5G
103812-572EA 8 USD
103812-572 103852-468 77193-860 77304-852 77304-854
(2-Aminoethoxy)benzene 98%
(2-Aminoethoxy)benzene

Precaución: for Research Use Only

Formula: C₆H₅OCH₂CH₂NH₂
MW: 137.18 g/mol
Flash Pt: 107 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00052975
CAS Number: 1758-46-9
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR