2-Pentylpyridine 97%

Supplier: Ambeed
Warning

Synonyms: 2-Amylpyridine

A133683-100G A133683-25G A133683-5G
77095-084EA 88 USD
77095-084 77314-582 77314-584
2-Pentylpyridine 97%
2-Pentylpyridine

Precaución: for Research Use Only

Formula: C₁₀H₁₅N
MW: 149.24 g/mol
Boiling Pt: 210.4 °C (760 mm Hg)
Flash Pt: 84.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00051828
CAS Number: 2294-76-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR