2-Pentyl-1H-benzo[d]imidazole 97%
Supplier: Ambeed
|
Warning
|
A270205-1G
A270205-5G
A270205-10G
103812-990EA
6
USD
103812-990
103852-896
77092-664
77092-666
77238-710
2-Pentyl-1H-benzo[d]imidazole 97%
2-Pentyl-1H-benzo[d]imidazole
Precaución: for Research Use Only
Formula:
C₁₂H₁₆N₂ Boiling Pt: 369 °C (760 mm Hg) Flash Pt: 184.5 °C Storage Temperature: Ambient |
MDL Number:
MFCD00628961 CAS Number: 5851-46-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...