2-Oxopiperidine-4-carboxylic acid 97%

Supplier: Ambeed
Warning

A154026-250MG A154026-1G A154026-5G
103804-596EA 36 USD
103804-596 103842-776 76973-244 77318-462 77318-464
2-Oxopiperidine-4-carboxylic acid 97%
2-Oxopiperidine-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₉NO₃
MW: 143.1400 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13659408
CAS Number: 24537-50-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR