2-Oxoindoline-4-carboxylic acid 96%

Supplier: Ambeed
Warning

A210504-250MG A210504-1G A210504-5G
103818-008EA 58 USD
103818-008 103848-704 76873-656 77087-074 77087-076
2-Oxoindoline-4-carboxylic acid 96%
2-Oxoindoline-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₇NO₃
MW: 177.1600 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10000702
CAS Number: 90322-37-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR