Phenylglyoxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: 2-Oxo-2-phenylacetic acid, Benzoylformic acid

A109032-100G A109032-25G A109032-500G A109032-5G
103823-220EA 23 USD
103823-220 77233-408 77233-410 77233-412 77283-800
Phenylglyoxylic acid 95%
Phenylglyoxylic acid

Precaución: for Research Use Only

Formula: C₈H₆O₃
MW: 150.13 g/mol
Boiling Pt: 258.6 °C (760 mm Hg)
Melting Pt: 60…69 °C
Flash Pt: 124.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00002575
CAS Number: 611-73-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR