2-Hydroxy-1H-imidazole-4-carboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: 2-Oxo-2,3-dihydro-1H-imidazole-4-carboxylic acid

A581005-1G A581005-5G A581005-100MG A581005-250MG
103828-864EA 195 USD
103828-864 103840-014 76844-146 76844-148 77169-932 77169-934
2-Hydroxy-1H-imidazole-4-carboxylic acid 95%
2-Hydroxy-1H-imidazole-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₄H₄N₂O₃
MW: 128.09 g/mol
Melting Pt: 270 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02323264
CAS Number: 39828-47-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR