2-(Hydroxymethyl)-1,3-oxazole 96%

Supplier: Ambeed
Warning

Synonyms: 2-Oxazolemethanol

A144653-100MG A144653-1G A144653-5G A144653-250MG
103803-432EA 70 USD
103803-432 103841-510 77272-648 77299-404 77299-406 77748-010
2-(Hydroxymethyl)-1,3-oxazole 96%
2-(Hydroxymethyl)-1,3-oxazole

Precaución: for Research Use Only

Formula: C₄H₅NO₂
MW: 99.09 g/mol
Boiling Pt: 174.6 °C (760 mm Hg)
Storage Temperature: Freezer
MDL Number: MFCD08703642
CAS Number: 130551-92-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR