2-Oxaspiro[3.3]heptan-6-ol 97%

Supplier: Ambeed
Warning

A229292-100MG A229292-250MG A229292-1G
103819-462EA 103 USD
103819-462 103850-362 77013-478 77013-480 77089-190
2-Oxaspiro[3.3]heptan-6-ol 97%
2-Oxaspiro[3.3]heptan-6-ol

Precaución: for Research Use Only

Formula: C₆H₁₀O₂
MW: 114.14 g/mol
Boiling Pt: 223.3±8.0 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD20926156
CAS Number: 1363381-08-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR