2-Oxa-7-azaspiro[4.5]decane hydrochloride 95%

Supplier: Ambeed
Warning

A181783-250MG A181783-1G A181783-100MG
103807-644EA 353 USD
103807-644 103846-018 76872-780 77301-742 77671-564
2-Oxa-7-azaspiro[4.5]decane hydrochloride 95%
2-Oxa-7-azaspiro[4.5]decane hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₆ClNO
MW: 177.67 g/mol
Flash Pt: 115.5 °C
Storage Temperature: Ambient
MDL Number: MFCD13185095
CAS Number: 374795-37-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR