2-Oxa-7-azaspiro[3.5]nonane hemioxalate 96%

Supplier: Ambeed
Warning

Synonyms: 2-Oxa-7-azaspiro[3.5]nonane oxalate(2:1)

A236185-100MG A236185-250MG A236185-5G
103811-074EA 95 USD
103811-074 76826-592 76826-594 76826-596
2-Oxa-7-azaspiro[3.5]nonane hemioxalate 96%
2-Oxa-7-azaspiro[3.5]nonane hemioxalate

Precaución: for Research Use Only

Formula: C₁₆H₂₈N₂O₆
MW: 344.4 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11035888
CAS Number: 1429056-28-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR