2-Oxa-6-azaspiro[3.3]heptane oxalate (2:1) 97%

Supplier: Ambeed
Warning

A494268-10G A494268-250MG A494268-1G A494268-25G A494268-5G A494268-100G
103828-238EA 11 USD
103828-238 103839-316 76818-694 76818-696 76843-278 76843-280 76843-282 77673-286
2-Oxa-6-azaspiro[3.3]heptane oxalate (2:1) 97%
2-Oxa-6-azaspiro[3.3]heptane oxalate (2:1)

Precaución: for Research Use Only

Formula: C₁₂H₂₀N₂O₆
Storage Temperature: Refrigerator
MDL Number: MFCD16652332
CAS Number: 1045709-32-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR