2-Oxa-6-azaspiro[3.3]heptane oxalate 97%
Supplier: Ambeed
|
Warning
|
A161934-250MG
A161934-1G
103805-554EA
16
USD
103805-554
103843-798
76973-624
77300-564
2-Oxa-6-azaspiro[3.3]heptane oxalate 97%
2-Oxa-6-azaspiro[3.3]heptane oxalate
Precaución: for Research Use Only
Formula:
C₇H₁₁NO₅ Storage Temperature: Freezer |
MDL Number:
MFCD11976178 CAS Number: 1159599-99-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...