2-Nitropyridine-5-carboxylic acid 97%
Supplier: Ambeed
|
Warning
|
A298654-25G
A298654-100MG
A298654-250MG
A298654-1G
A298654-5G
103814-190EA
21
USD
103814-190
103835-336
76838-036
76977-690
76977-692
77305-746
77305-748
2-Nitropyridine-5-carboxylic acid 97%
2-Nitropyridine-5-carboxylic acid
Precaución: For research use only
Formula:
C₆H₄N₂O₄ MW: 168.1070 g/mol Boiling Pt: 434.9 °C (760 mm Hg) Melting Pt: 183 °C Flash Pt: 216.8 °C Storage Temperature: Ambient |
MDL Number:
MFCD04114183 CAS Number: 33225-73-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...