2-Nitroisophthalic acid 98%

Supplier: Ambeed
Warning

A177308-100G A177308-10G A177308-25G A177308-500G A177308-1G A177308-5G
103807-224EA 5 USD
103807-224 103845-552 77273-906 77273-908 77273-910 77273-912 77301-498 77301-500
2-Nitroisophthalic acid 98%
2-Nitroisophthalic acid

Precaución: for Research Use Only

Formula: C₈H₅NO₆
MW: 211.1284 g/mol
Boiling Pt: 473.7 °C (760 mm Hg)
Melting Pt: 314 °C
Flash Pt: 214 °C
Storage Temperature: Ambient
MDL Number: MFCD00017007
CAS Number: 21161-11-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR