2-Nitrobenzyl alcohol 98%

Supplier: Ambeed
Warning

Synonyms: (2-Nitrophenyl)methanol

A208767-1000G A208767-25G A208767-5G A208767-100G A208767-10G A208767-500G
103809-442EA 159 USD
103809-442 103848-570 77086-896 77086-898 77086-900 77275-086 77275-088 77275-090
2-Nitrobenzyl alcohol 98%
2-Nitrobenzyl alcohol

Precaución: for Research Use Only

Formula: C₇H₇NO₃
MW: 153.14 g/mol
Boiling Pt: 270 °C (760 mm Hg)
Melting Pt: 69…72 °C
Storage Temperature: Ambient
MDL Number: MFCD00007186
CAS Number: 612-25-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR