2-Nitrobenzoic acid 98%

Supplier: Ambeed
Warning

A360419-1000G A360419-25G A360419-5G A360419-100G A360419-500G
103810-216EA 188 USD
103810-216 103837-262 76840-466 76840-468 76840-470 77278-296 77278-298
2-Nitrobenzoic acid 98%
2-Nitrobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₅NO₄
Melting Pt: 146…148 °C
Storage Temperature: Ambient
MDL Number: MFCD00007137
CAS Number: 552-16-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR