2-Nitro-5-(trifluoromethyl)benzenol 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Nitro-5-(trifluoromethyl)phenol, 3-Hydroxy-4-nitrobenzotrifluoride
A397875-1G
A397875-25G
A397875-5G
A397875-250MG
103827-006EA
35
USD
103827-006
103838-046
76841-480
76841-482
76841-484
77003-854
2-Nitro-5-(trifluoromethyl)benzenol 95%
2-Nitro-5-(trifluoromethyl)benzenol
Precaución: for Research Use Only
Formula:
C₇H₄F₃NO₃ MW: 207.11 g/mol Boiling Pt: 239.355 °C (760 mm Hg) Flash Pt: 98.559 °C Storage Temperature: Ambient |
MDL Number:
MFCD08282785 CAS Number: 402-17-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...