2-Nitro-5-(trifluoromethyl)benzaldehyde 97%

Supplier: Ambeed
Warning

A437171-1G A437171-250MG
103827-620EA 95 USD
103827-620 103838-686 76842-424 77890-893
2-Nitro-5-(trifluoromethyl)benzaldehyde 97%
2-Nitro-5-(trifluoromethyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₈H₄F₃NO₃
Storage Temperature: Refrigerator
MDL Number: MFCD13185768
CAS Number: 1176723-57-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR