2-Nitro-4-(trifluoromethyl)phenol 98%

Supplier: Ambeed
Danger

Synonyms: 4-Hydroxy-3-nitrobenzotrifluoride

A111331-100G A111331-25G A111331-1G A111331-5G
103797-820EA 7 USD
103797-820 103823-636 76765-830 76765-832 77310-440 77310-442
2-Nitro-4-(trifluoromethyl)phenol 98%
2-Nitro-4-(trifluoromethyl)phenol

Precaución: for Research Use Only

Formula: C₇H₄F₃NO₃
MW: 207.11 g/mol
Boiling Pt: 207.4 °C (760 mm Hg)
Flash Pt: 95 °C
Storage Temperature: Ambient
MDL Number: MFCD00009791
CAS Number: 400-99-7
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR