2-Nitro-1-naphthol 98%

Supplier: Ambeed
Warning

Synonyms: 1-Hydroxy-2-nitronaphthalene

A153498-100G A153498-10G A153498-1G A153498-25G A153498-5G
103804-520EA 6 USD
103804-520 103842-694 76798-734 76798-736 77300-008 77318-354 77318-356
2-Nitro-1-naphthol 98%
2-Nitro-1-naphthol

Precaución: for Research Use Only

Formula: C₁₀H₇NO₃
Boiling Pt: 336.7 °C (760 mm Hg)
Melting Pt: 123…125 °C
Flash Pt: 149.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00003956
CAS Number: 607-24-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR