2-Nitro-1H-pyrrole 95%

Supplier: Ambeed
Warning

A562079-1G A562079-5G A562079-10G A562079-250MG
103828-746EA 68 USD
103828-746 103839-880 76843-980 76843-982 77034-916 77034-918
2-Nitro-1H-pyrrole 95%
2-Nitro-1H-pyrrole

Precaución: for Research Use Only

Formula: C₄H₄N₂O₂
MW: 112.087 g/mol
Melting Pt: 63…67 °C
Flash Pt: 115.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD12923841
CAS Number: 5919-26-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR