2-Naphthylacetonitrile 97%

Supplier: Ambeed
Warning

A226350-100G A226350-10G A226350-25G A226350-1G A226350-5G
103819-242EA 6 USD
103819-242 103850-144 76722-118 76722-120 77088-874 77303-768 77303-770
2-Naphthylacetonitrile 97%
2-Naphthylacetonitrile

Precaución: for Research Use Only

Formula: C₁₂H₉N
MW: 167.21 g/mol
Melting Pt: 82…84 °C
Flash Pt: 303 °C
Storage Temperature: Ambient
MDL Number: MFCD00004122
CAS Number: 7498-57-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR