2-Naphthalenemethanol 98%

Supplier: Ambeed
Warning

A294730-25G A294730-5G A294730-100G A294730-10G A294730-500G
103835-162EA 7 USD
103835-162 76837-802 76837-804 77002-386 77002-388 77002-390
2-Naphthalenemethanol 98%
2-Naphthalenemethanol

Precaución: for Research Use Only

Formula: C₁₁H₁₀O
MW: 158.2000 g/mol
Boiling Pt: 319.6 °C (760 mm Hg)
Melting Pt: 79…81 °C
Flash Pt: 154 °C
Storage Temperature: Ambient
MDL Number: MFCD00004124
CAS Number: 1592-38-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR