2-Naphthyl boronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-Naphthaleneboronic acid, Naphthalene-2-boronic acid, 2-Naphthyl boric acid

A195894-100G A195894-10G A195894-500G A195894-25G A195894-1G A195894-5G
103816-792EA 8 USD
103816-792 103847-406 76769-968 76769-970 76769-972 77085-354 77302-484 77302-486
2-Naphthyl boronic acid 98%
2-Naphthyl boronic acid

Precaución: for Research Use Only

Formula: C₁₀H₉BO₂
MW: 171.99 g/mol
Boiling Pt: 381.9 °C (760 mm Hg)
Melting Pt: 269…275 °C
Storage Temperature: Ambient
MDL Number: MFCD00236051
CAS Number: 32316-92-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR