N-(2-Hydroxyethyl)morpholine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-(2-Hydroxyethyl)morpholine, 2-Morpholinoethanol
A193705-1000G
A193705-100G
A193705-500G
A193705-25G
103847-184EA
5
USD
103847-184
76721-200
76721-202
76721-204
77302-356
N-(2-Hydroxyethyl)morpholine 97%
N-(2-Hydroxyethyl)morpholine
Precaución: for Research Use Only
Formula:
C₆H₁₃NO₂ MW: 131.17 g/mol Boiling Pt: 224.4 °C (760 mm Hg) Melting Pt: 1 °C Flash Pt: 98 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006180 CAS Number: 622-40-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...