2-Morpholinobenzaldehyde 97%

Supplier: Ambeed
Warning

A352413-10G A352413-250MG A352413-1G A352413-25G A352413-5G
103826-068EA 20 USD
103826-068 103837-068 76803-310 76803-312 76840-176 76840-178 76840-180
2-Morpholinobenzaldehyde 97%
2-Morpholinobenzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₃NO₂
MW: 191.2300 g/mol
Boiling Pt: 342.8 °C (760 mm Hg)
Melting Pt: 45…48 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00662562
CAS Number: 58028-76-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR