2-Morpholino-5-(1H-pyrrol-1-yl)benzoic acid 97%

Supplier: Ambeed
Warning

A301474-100MG A301474-250MG
103814-306EA 1145 USD
103814-306 103835-458 77277-340 77277-342
2-Morpholino-5-(1H-pyrrol-1-yl)benzoic acid 97%
2-Morpholino-5-(1H-pyrrol-1-yl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₆N₂O₃
MW: 272.29 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03406724
CAS Number: 690632-76-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR