2-Methylthiophene-3-carboxylic acid 98%

Supplier: Ambeed
Warning

A142357-10G A142357-1G A142357-5G
103803-148EA 12 USD
103803-148 77272-530 77316-372 77316-374
2-Methylthiophene-3-carboxylic acid 98%
2-Methylthiophene-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₆O₂S
Flash Pt: 118 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01859891
CAS Number: 1918-78-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR