2-Methylthiazole-5-carbaldehyde 97%

Supplier: Ambeed
Warning

A129997-10G A129997-1G A129997-25G A129997-5G
103801-360EA 12 USD
103801-360 76782-298 77313-776 77313-778 77313-780
2-Methylthiazole-5-carbaldehyde 97%
2-Methylthiazole-5-carbaldehyde

Precaución: for Research Use Only

Formula: C₅H₅NOS
MW: 127.1600 g/mol
Boiling Pt: 219 °C (760 mm Hg)
Melting Pt: 40…44 °C
Flash Pt: 86.2 °C
Storage Temperature: Freezer
MDL Number: MFCD09864617
CAS Number: 1003-60-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR