2-Methyl-4-thiazolecarboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-Methylthiazole-4-carboxylic acid

A204875-1G A204875-25G A204875-5G A204875-100G A204875-10G A204875-250MG
103817-574EA 15 USD
103817-574 103848-238 77086-426 77086-428 77086-430 77164-720 77164-722 77164-724
2-Methyl-4-thiazolecarboxylic acid 97%
2-Methyl-4-thiazolecarboxylic acid

Precaución: for Research Use Only

Formula: C₅H₅NO₂S
MW: 143.1637 g/mol
Boiling Pt: 301.2 °C (760 mm Hg)
Melting Pt: 148 °C
Storage Temperature: Ambient
MDL Number: MFCD03407332
CAS Number: 35272-15-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR