2-Methylthiazole 98%

Supplier: Ambeed
Danger

A563595-25G A563595-5G A563595-100G A563595-10G A563595-1G
103839-896EA 11 USD
103839-896 76844-002 76844-004 77292-554 77292-556 77292-558
2-Methylthiazole 98%
2-Methylthiazole

Precaución: for Research Use Only

Formula: C₄H₅NS
MW: 99.16 g/mol
Boiling Pt: 128.5 °C (760 mm Hg)
Melting Pt: 24 °C
Storage Temperature: Ambient
MDL Number: MFCD00053144
CAS Number: 3581-87-1
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR