2-Methylthiazol-5-amine 95%

Supplier: Ambeed
Warning

A253042-100MG A253042-250MG A253042-1G A253042-5G
103812-072EA 97 USD
103812-072 103851-920 76874-666 76874-668 77091-346 77304-596
2-Methylthiazol-5-amine 95%
2-Methylthiazol-5-amine

Precaución: for Research Use Only

Formula: C₄H₆N₂S
MW: 114.17 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11847087
CAS Number: 89281-44-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR