2-Methylsulfanylpyrimidine-4-carboxylic acid 95%

Supplier: Ambeed
Warning

A208089-100G A208089-250MG A208089-25G A208089-1G A208089-5G
103817-854EA 17 USD
103817-854 103848-540 77112-360 77112-362 77112-364 77303-058 77303-060
2-Methylsulfanylpyrimidine-4-carboxylic acid 95%
2-Methylsulfanylpyrimidine-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₆N₂O₂S
Boiling Pt: 378.4 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD09834808
CAS Number: 1126-44-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR