2-Methylsulfanylpyrimidine-4-carbaldehyde 97%

Supplier: Ambeed
Warning

Synonyms: 2-(Methylthio)pyrimidine-4-carbaldehyde

A182346-1G A182346-5G A182346-10G A182346-250MG
103807-698EA 26 USD
103807-698 103846-068 77083-610 77083-612 77111-454 77111-456
2-Methylsulfanylpyrimidine-4-carbaldehyde 97%
2-Methylsulfanylpyrimidine-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₆H₆N₂OS
MW: 154.19 g/mol
Boiling Pt: 301.2 °C (760 mm Hg)
Melting Pt: 68 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02683211
CAS Number: 1074-68-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR