8-Amino-2-methylquinoline 98%

Supplier: Ambeed
Warning

Synonyms: 2-Methylquinolin-8-amine, 8-Aminoquinaldine

A238027-10G A238027-1G A238027-25G A238027-5G A238027-100G
103811-198EA 11 USD
103811-198 103850-982 76647-668 77090-084 77090-086 77090-088 77615-964
8-Amino-2-methylquinoline 98%
8-Amino-2-methylquinoline

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂
MW: 158.1998 g/mol
Boiling Pt: 316.6 °C (760 mm Hg)
Melting Pt: 56…58 °C
Flash Pt: 171.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00023998
CAS Number: 18978-78-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR