2-methylpyrimidine-5-carboxylic acid 97%
Supplier: Ambeed
|
Warning
|
A204856-1G
A204856-25G
A204856-5G
A204856-100G
A204856-100MG
A204856-10G
A204856-250MG
103817-572EA
8
USD
103817-572
103848-236
77086-420
77086-422
77086-424
77274-970
77274-972
77274-974
77274-976
2-methylpyrimidine-5-carboxylic acid 97%
2-methylpyrimidine-5-carboxylic acid
Precaución: for Research Use Only
Formula:
C₆H₆N₂O₂ Boiling Pt: 292.5 °C (760 mm Hg) Melting Pt: 203…205 °C Flash Pt: 130.706 °C Storage Temperature: Ambient |
MDL Number:
MFCD08235210 CAS Number: 5194-32-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...