2-Methylpyrimidin-5-ol 98%

Supplier: Ambeed
Warning

A175594-100MG A175594-250MG A175594-1G
103807-064EA 23 USD
103807-064 103845-376 76769-346 76769-348 77082-718
2-Methylpyrimidin-5-ol 98%
2-Methylpyrimidin-5-ol

Precaución: for Research Use Only

Formula: C₅H₆N₂O
MW: 110.1139 g/mol
Boiling Pt: 208 °C (760 mm Hg)
Melting Pt: 175…179 °C
Flash Pt: 80 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09879706
CAS Number: 35231-56-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR