2-Methylpyridine-3,4-diamine 95%

Supplier: Ambeed
Warning

A129127-100MG A129127-250MG
103801-230EA 279 USD
103801-230 77271-906 77271-908
2-Methylpyridine-3,4-diamine 95%
2-Methylpyridine-3,4-diamine

Precaución: for Research Use Only

Formula: C₆H₉N₃
MW: 123.16 g/mol
Flash Pt: 185.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD10000599
CAS Number: 15931-19-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR