2-Methylpyridin-3-ol 98%

Supplier: Ambeed
Warning

Synonyms: 3-Hydroxy-2-methylpyridine

A229033-1G A229033-5G A229033-500G A229033-10G A229033-100G
103819-438EA 8 USD
103819-438 103850-338 77089-156 77089-158 77112-910 77612-230 77685-622
2-Methylpyridin-3-ol 98%
2-Methylpyridin-3-ol

Precaución: for Research Use Only

Formula: C₆H₇NO
MW: 109.13 g/mol
Melting Pt: 168…169 °C
Flash Pt: 127.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00082538
CAS Number: 1121-25-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR