3-Amino-2-methylpyridine 98%

Supplier: Ambeed
Warning

Synonyms: 2-Methylpyridin-3-amine, 2-Methyl-3-pyridylamine, 3-Amino-2-picoline

A106022-100G A106022-10G A106022-1G A106022-25G A106022-5G
103822-738EA 22 USD
103822-738 77107-640 77107-642 77107-644 77107-646 77309-602
3-Amino-2-methylpyridine 98%
3-Amino-2-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₈N₂
MW: 108.14 g/mol
Boiling Pt: 236.8 °C (760 mm Hg)
Melting Pt: 114…119 °C
Flash Pt: 120.1 °C
Storage Temperature: Ambient
MDL Number: MFCD03788195
CAS Number: 3430-10-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR