2-Methylpyrazolo[1,5-a]pyrimidine-6-carboxylic acid 97%

Supplier: Ambeed
Warning

A109125-10G A109125-250MG A109125-1G A109125-5G A109125-25G
103797-450EA 9 USD
103797-450 103823-240 77160-622 77160-624 77310-058 77310-060 77683-002
2-Methylpyrazolo[1,5-a]pyrimidine-6-carboxylic acid 97%
2-Methylpyrazolo[1,5-a]pyrimidine-6-carboxylic acid

Precaución: For research use only.

Formula: C₈H₇N₃O₂
Melting Pt: 253…257 °C
Storage Temperature: Ambient
MDL Number: MFCD19443280
CAS Number: 739364-95-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR