2-Methylpropane-1,2-diol 98%

Supplier: Ambeed
Warning

Synonyms: Dimethyl glyoxal

A135285-100G A135285-10G A135285-1G A135285-25G A135285-5G
103802-126EA 25 USD
103802-126 76972-316 76972-318 77314-928 77314-930 77314-932
2-Methylpropane-1,2-diol 98%
2-Methylpropane-1,2-diol

Precaución: for Research Use Only

Formula: C₄H₁₀O₂
MW: 90.121 g/mol
Boiling Pt: 176 °C (760 mm Hg)
Flash Pt: 73.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00971926
CAS Number: 558-43-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR