2-Methylnicotinic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-Methylpyridine-3-carboxylic acid, 2-Methyl-3-pyridinecarboxylic acid

A156076-10G A156076-1G A156076-25G A156076-100G
103804-826EA 9 USD
103804-826 103843-030 77095-990 77318-768 77318-770 77685-284
2-Methylnicotinic acid 98%
2-Methylnicotinic acid

Precaución: for Research Use Only

Formula: C₇H₇NO₂
Boiling Pt: 280.4 °C (760 mm Hg)
Melting Pt: 228…230 °C
Flash Pt: 123.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00013449
CAS Number: 3222-56-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR