3-Formyl-2-picoline 97%

Supplier: Ambeed
Warning

Synonyms: 2-Methylnicotinaldehyde

A179273-1G A179273-5G A179273-100MG A179273-250MG
103807-412EA 44 USD
103807-412 103845-758 77083-216 77083-218 77273-978 77273-980
3-Formyl-2-picoline 97%
3-Formyl-2-picoline

Precaución: for Research Use Only

Formula: C₇H₇NO
MW: 121.14 g/mol
Flash Pt: 93.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08234911
CAS Number: 60032-57-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR